Learn More
DSI LLC Backpressure Regulator, variable 1300 - 4200psi, max. 80C, complete, for LC and SFC
Catalog No. 502255022
Encompass_Preferred
Backpressure Regulator, variable 1300 - 4200psi, max. 80C, complete, for LC and SFC
Catalog No. 50-225-5022 Supplier DSI LLC Supplier No. JRBPR3
Backpressure Regulator, variable 1300 - 4200psi, max. 80C, complete, for LC and SFC