Learn More
Electron Microscopy Sciences MAC Backscattered Electron Calibration Standard
SDP
Supplier:  Electron Microscopy Sciences 80070BED
Encompass_Preferred
30 mm x 5 mm carbon block that checks detector and SEM performance. Certified standards for system calibration. Contains standards to check 1 atomic number resolution. Faraday cup to set probe current. Duplex brass to check 0.1 atomic number resolution.
Catalog No. 50-190-1613