Learn More
Med Vet International Disposable Underpad Attends Care Dri-Sorb Advanced 30 X 30 Inch Cellulose / Polymer Heavy Absorbency, UFP-300, 10/Pack
Catalog No. 503043080
Encompass_Preferred
Attends Dri-Sorb underpads have a cloth-like top sheet with excellent wicking properties to keep the skin dry and protected The core contains cellulose fiber and a super-absorbent polymer for effective results These 30-inch x 30-inch peach nursing underpads come with a back sheet that is leakproof to protect linens and surfaces They're ideal for everyday use There are 10 Attends disposable underpads in a bag
Catalog No. 50-304-3080 Supplier Med Vet International Supplier No. 953632PK
Attends Dri-Sorb underpads have a cloth-like top sheet with excellent wicking properties to keep the skin dry and protected The core contains cellulose fiber and a super-absorbent polymer for effective results These 30-inch x 30-inch peach nursing underpads come with a back sheet that is leakproof to protect linens and surfaces They're ideal for everyday use There are 10 Attends disposable underpads in a bag