Learn More
Electron Microscopy Sciences Omniprobe Short-Cut™ Coupons Backside Thinning, Copper, 26.5° Angle
SDP
Catalog No. 5033779
Encompass
TEM sample grid coupon designed specially for use in the Short-Cut™ tool. The center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut.
Material: Copper
Angle: 26.5°
Catalog No. 50-337-79 Supplier Electron Microscopy Sciences Supplier No. 75977CU
TEM sample grid coupon designed specially for use in the Short-Cut™ tool. The center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut.
Material: Copper
Angle: 26.5°