Learn More
Electron Microscopy Sciences Omniprobe Short-Cut™ Coupons Backside Thinning, Copper, 45° Angle
SDP
Catalog No. 5033781
Encompass
TEM sample grid coupon designed specially for use in the Short-Cut™ tool. The center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut.
Material: Copper
Angle: 45°
Catalog No. 50-337-81 Supplier Electron Microscopy Sciences Supplier No. 75975CU
TEM sample grid coupon designed specially for use in the Short-Cut™ tool. The center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut.
Material: Copper
Angle: 45°