Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Backyard Bugs™
SCI_ED

Catalog No. S78997
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S78997 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Catalog No. S78997 Supplier Learning Resources, Inc Supplier No. LER 0457
Add to Cart
Add to Cart

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Includes

  • Bugs: Grasshopper, bumblebee, beetle, spider, dragonfly, and caterpillar
  • Reusable plastic tub
  • Suggested activities

A bucket of bugs.

Creepy, crawly critters are great for sorting and counting activities. Soft rubber bugs come in six colors and measure approximately 2 in. (5cm).

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Backyard Bugs™ Set
Includes Bugs, grasshopper, bumblebee, beetle, spider, dragonfly and caterpillar, reusable Plastic tub, suggested activities
Quantity Set of 72
For Use With (Application) These colorful creepy, crawly counters turn sorting and counting activities into fun adventures
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.