Learn More
Backyard Pets

Description
- Find out how to catch and care for lots of different, easy-to-find critters
- Find out how to attract birds and butterflies to homemade feeders and gardens
- Learn how to interpret fireflies' signals
- Learn how to entice worms from their burrows with sound
- Discover how they see, hear, and move, and even how they help the environment
- Carol A. Amato; Paperback; 2002; 128 pages
Specifications
Specifications
| Format | Paperback Book |
| Product Type | Backyard Pets |
| For Use With (Application) | Activities for exploring wildlife close to home |
| Subject | Biology |
| Author | Carol A. Amato |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.