Learn More
Description
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
- 10 -6spore population
Specifications
Specifications
| ATCC® | 7953TM* |
| Description | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population |
| Format | Spore Strips |
| Product Type | Quality Control Organism |
| Target Organism Class | Gram-positive Rods |
| Test Type | Qualitative QC |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.