Learn More
Apexbio Technology LLC (±)-Baclofen 1134-47-0 10g
SDP
Catalog No. 5000089133
Encompass_Preferred
( )-Baclofen (CAS 1134-47-0) is a selective agonist of the -aminobutyric acid type B (GABAB) receptor a G protein-coupled receptor involved in inhibitory neurotransmission Exhibiting an IC50 of 200 nM ( )-baclofen induces skeletal muscle relaxation and reduces spasticity by activating GABAB-mediated signaling pathways In cellular studies ( )-baclofen inhibits the proliferation of human hepatocellular carcinoma (HCC) cells in a dose-dependent manner induces G0/G1 cell cycle arrest and modulates intracellular cAMP and p21WAF1 expression In vivo it suppresses tumor growth in HCC xenograft models without evident toxicity ( )-Baclofen is thus valuable for investigating GABAB-mediated mechanisms in cancer and neuropharmacology
Catalog No. 50-000-89133 Supplier Apexbio Technology LLC Supplier No. C405110000
( )-Baclofen (CAS 1134-47-0) is a selective agonist of the -aminobutyric acid type B (GABAB) receptor a G protein-coupled receptor involved in inhibitory neurotransmission Exhibiting an IC50 of 200 nM ( )-baclofen induces skeletal muscle relaxation and reduces spasticity by activating GABAB-mediated signaling pathways In cellular studies ( )-baclofen inhibits the proliferation of human hepatocellular carcinoma (HCC) cells in a dose-dependent manner induces G0/G1 cell cycle arrest and modulates intracellular cAMP and p21WAF1 expression In vivo it suppresses tumor growth in HCC xenograft models without evident toxicity ( )-Baclofen is thus valuable for investigating GABAB-mediated mechanisms in cancer and neuropharmacology