Learn More
Cayman Chemical (±)-Baclofen-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1189938-30-4, 1 mg
SDP
Catalog No. 502938443
Encompass_Preferred
An internal standard for the quantification of (±)-baclofen by GC- or LC-MS
Catalog No. 50-293-8443 Supplier Cayman Chemical Supplier No. 278111 MG
An internal standard for the quantification of (±)-baclofen by GC- or LC-MS