Learn More
Medchemexpress LLC Baclofen hydrochloride | 28311-31-1 | 99.78% | C10H13Cl2NO2 | 5 MG
Supplier: Medchemexpress LLC HYB0007C5MG
Baclofen hydrochloride is a lipophilic derivative of γ-aminobutyric acid (GABA) and acts as an orally active, selective metabotropic GABAB receptor (GABABR) agonist. It mimics the action of GABA, producing slow presynaptic inhibition via the GABAB receptor. It exhibits high blood-brain barrier penetrance and is considered for muscle spasticity research.
- Orally active, selective GABABR agonist
- Mimics GABA action
- Produces slow presynaptic inhibition
- High blood-brain barrier penetrance
- Potential for muscle spasticity research