Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baclofen, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600840 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-840 5 g Amber Glass Bottle
18-600-841 25 g Poly Bottle
18-600-843 100 g Poly Bottle
18-600-844 500 g Amber Glass Bottle
18-600-842 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-840 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B10495GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

B1049, 1134-47-0, C10H12ClNO2

Spectrum™ Chemical Baclofen, USP is a muscle relaxer and an antispastic agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1134-47-0
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.66
InChI Key KPYSYYIEGFHWSV-UHFFFAOYNA-N
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
SMILES NCC(CC(O)=O)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Packaging Amber Glass Bottle
UN Number UN2811
Quantity 5 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.