Learn More
Cayman Chemical -BaclofenhydrochlorIde 10mg
SDP
Catalog No. 502991229
Encompass_Preferred
A GABAB receptor agonist; increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg; reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg
Catalog No. 50-299-1229 Supplier Cayman Chemical Supplier No. 3159510 MG
A GABAB receptor agonist; increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg; reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg