Learn More
Cayman Chemical (+)-Baclofen (hydrochloride), Purity: ≥98%, CAS: 63701-55-3, 50 mg
SDP
Catalog No. 502988021
Encompass_Preferred
A GABAB receptor agonist; increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg; reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg
Catalog No. 50-298-8021 Supplier Cayman Chemical Supplier No. 3159550 MG
A GABAB receptor agonist; increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg; reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg