Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BacMam pCMV-Dest Vector

Catalog No. A24223
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24223 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24223 Supplier Invitrogen™ Supplier No. A24223
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The BacMam pCMV-Dest Vector combines Gateway™ cloning and BacMam gene expression technologies for easy recombination-based cloning and baculovirus-based expression of a target gene in variety of cell types.

The BacMam pCMV-Dest Vector combines Gateway™ cloning and BacMam gene expression technologies for easy recombination-based cloning and baculovirus-based expression of a target gene in variety of cell types. With BacMam technology, a modified insect cell virus (baculovirus) is used as a vehicle to efficiently deliver and transiently express genes in mammalian cells with minimum effort and toxicity. This technology allows for control over the level of expression through the increase or decrease of viral particle concentration. When combined with Gateway™ cloning technology, a variety of sizes of target genes can be cloned and expressed. Sizes ranging from an insert for RNAi to a large gene with introns (38 kb) have been cloned and expressed successfully using the BacMam pCMV-Dest Vector. This plasmid accommodates various cloning schemes, including Gateway™, Seamless Assembly, and traditional cloning using restriction enzymes.

The BacMam pCMV-Dest Vector contains a CMV promoter to drive constitutive expression of the target gene. It also contains VSV-G elements, which enable baculovirus to have high transduction efficiency, and a WPRE element, which elongates transient expression in mammalian systems. To facilitate successful cloning, AmpR and gentamicin have been incorporated for positive selection, as well as Cm(R) (chloramphenicol resistance gene) for negative selection. Upon successful cloning, the region containing the CmR is replaced by the insert, selecting against any bacteria that still have a CmR-containing region.

TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Baculovirus (BacMam System)
Promoter CMV
Product Type Insect Cell Expression Vector
Reporter Gene None
Selection Agent (Eukaryotic) None
Content And Storage 10 μg of BacMam pCMV-Dest Vector.
Store at 2°C to 8°C.
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS, Seamless Cloning, Gateway
Concentration 500 ng/μL
Quantity 10 μg
Vector pDEST
Product Line ViraPower
For Use With (Application) Viral Expression, Constitutive Expression, Multi-Gene Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.