Learn More
MOLTOX Molecular Toxicology Inc Bacmid Selection, 30 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948625
Encompass_Preferred
LB Agar, Lennox plates supplemented with Gentamicin, Kanamycin, Tetracycline, Bluo-gal and IPTG used for transformation of recombinant Bacmid DNA. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-625 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S125
LB Agar, Lennox plates supplemented with Gentamicin, Kanamycin, Tetracycline, Bluo-gal and IPTG used for transformation of recombinant Bacmid DNA. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.