Learn More
Medchemexpress LLC HY-N6015 5mg Medchemexpress, Bacopasaponin C CAS:178064-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60155MG
Encompass_Preferred
Medchemexpress, HY-N6015 5mg Bacopasaponin C CAS:178064-13-6 Bacopasaponin C is an indigenous glycoside isolated from Bacopa monniera, with antitumor and anti-leishmanial activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0444