Learn More
Cayman Chemical Bacopaside II, Purity: ≥95%, CAS: 382146-66-9, 10 mg
SDP
Catalog No. 502954986
Encompass_Preferred
A triterpene glycoside; decreases hydrogen peroxide-induced intracellular ROS production and cell death in N2a neuroblastoma cells at 0.4 mg/ml; decreases immobility time in the forced swim and tail suspension tests in mice, indicating anti-depressant like activity, at 50 mg/kg; decreases migration and tube formation in 2H11 and 3B11 cells, as well as HUVECs when used at greater than 15 µM; inhibits the growth of MDA-MB-231, SHG-44, HCT8, A549, and PC3M cancer cells (IC50s = 32.4, 36.9, 40.3, 44.4, and 45.4 µM, respectively)
Catalog No. 50-295-4986 Supplier Cayman Chemical Supplier No. 1182210 MG
A triterpene glycoside; decreases hydrogen peroxide-induced intracellular ROS production and cell death in N2a neuroblastoma cells at 0.4 mg/ml; decreases immobility time in the forced swim and tail suspension tests in mice, indicating anti-depressant like activity, at 50 mg/kg; decreases migration and tube formation in 2H11 and 3B11 cells, as well as HUVECs when used at greater than 15 µM; inhibits the growth of MDA-MB-231, SHG-44, HCT8, A549, and PC3M cancer cells (IC50s = 32.4, 36.9, 40.3, 44.4, and 45.4 µM, respectively)