Learn More
Medchemexpress LLC HY-N6016 1mg Medchemexpress, Bacopaside II CAS:382146-66-9 Purity:>98%
SDP
Catalog No. 501970445
Encompass_Preferred
Medchemexpress, HY-N6016 1mg Bacopaside II CAS:382146-66-9 Bacopaside II, an extract from the medicinal herb Bacopa monnieri, blocks the Aquaporin-1 (AQP1) water channel and impairs migration of cells that express AQP1. Bacopaside II induces cell cycle arrest and apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0445 Supplier Medchemexpress LLC Supplier No. HYN60161MG
Medchemexpress, HY-N6016 1mg Bacopaside II CAS:382146-66-9 Bacopaside II, an extract from the medicinal herb Bacopa monnieri, blocks the Aquaporin-1 (AQP1) water channel and impairs migration of cells that express AQP1. Bacopaside II induces cell cycle arrest and apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.