Learn More
Medchemexpress LLC Bacopaside II | 382146-66-9 | MFCD32182525 | 98.7% | 929.10 g·mol⁻1 | C47H76O18 | 10 MG
SDP
Catalog No. 5000289824
Encompass_Preferred
Bacopaside II is a triterpene glycoside isolated from Bacopa monnieri, supplied for research use. It has reported aquaporin-1 (AQP1) inhibitory activity and has demonstrated anti-angiogenesis and pro-apoptotic effects in cellular studies. Offered as a solid powder and as a DMSO solution for research applications.
  • Research-grade triterpene glycoside with reported AQP1 inhibitory activity.
  • Exhibits anti-angiogenesis activity and induces apoptosis in cell models.
  • High purity: 98.69% by manufacturer analysis.
  • Available as solid powder and as a 10 mM solution in DMSO for convenience.
  • Suitable for biochemical, cellular, and pharmacological research applications.
Catalog No. 50-002-89824 Supplier Medchemexpress LLC Supplier No. HYN601610MG
Bacopaside II is a triterpene glycoside isolated from Bacopa monnieri, supplied for research use. It has reported aquaporin-1 (AQP1) inhibitory activity and has demonstrated anti-angiogenesis and pro-apoptotic effects in cellular studies. Offered as a solid powder and as a DMSO solution for research applications.
  • Research-grade triterpene glycoside with reported AQP1 inhibitory activity.
  • Exhibits anti-angiogenesis activity and induces apoptosis in cell models.
  • High purity: 98.69% by manufacturer analysis.
  • Available as solid powder and as a 10 mM solution in DMSO for convenience.
  • Suitable for biochemical, cellular, and pharmacological research applications.