Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacopaside II, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014097 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4097 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4097 Supplier MilliporeSigma Supelco Supplier No. 446985MG
Add to Cart
Add to Cart

Bacopaside II is an antidepressant, belonging to the class of triterpenoid saponins isolated from the plant, Bacopa monnieri. Its potential applications include the improvement of high order cognitive processes and treatment/prevention of neurological diseases.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 382146-66-9
Physical Form Neat
Linear Formula C47H76O18
Synonym Pseudojujubogenin 3-O-[alpha-L-arabinofuranosyl-(1->2)-[beta-D-glucopyranosyl-(1->3)]-beta-D-glucopyranoside]
Type Analytical Standard
Quantity 5 mg
Formula Weight 929.10
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.