Learn More
Cayman Chemical BacopasIde X 10mg
SDP
Catalog No. 502953334
Encompass_Preferred
A triterpenoid saponin; inhibits the growth of C. albicans, C. neoformans, and A. fumigatus (MIC = 50 UG/ml for all); decreases the proliferation of various cancer cell lines (IC50s = 9.7-15.9 µM); reduces tumor growth by 84.13% in an S180 sarcoma mouse allograft model at 50 µmol/kg
Catalog No. 50-295-3334 Supplier Cayman Chemical Supplier No. 1185110 MG
A triterpenoid saponin; inhibits the growth of C. albicans, C. neoformans, and A. fumigatus (MIC = 50 UG/ml for all); decreases the proliferation of various cancer cell lines (IC50s = 9.7-15.9 µM); reduces tumor growth by 84.13% in an S180 sarcoma mouse allograft model at 50 µmol/kg