Learn More
Medchemexpress LLC Bacopaside X | 94443-88-6 | MFCD09752776 | 98.0% | 899.07 | C46H74O17 | 10 MG
SDP
Catalog No. 5000289832
Encompass_Preferred
Bacopaside X is a triterpenoid saponin isolated from Bacopa monnieri with reported binding affinity for the dopamine D1 receptor. It is supplied as a solid analytical material intended for research use only and is used in receptor binding and in vitro pharmacology studies.
  • naturally derived saponin suitable for biochemical assays
  • reported D1 receptor binding for receptor pharmacology research
  • available in small, weighed quantities for analytical work
  • characterized by established identifiers such as CAS number for unambiguous referencing
  • intended for research use only; not for human or clinical use
Catalog No. 50-002-89832 Supplier Medchemexpress LLC Supplier No. HYN514010MG
Bacopaside X is a triterpenoid saponin isolated from Bacopa monnieri with reported binding affinity for the dopamine D1 receptor. It is supplied as a solid analytical material intended for research use only and is used in receptor binding and in vitro pharmacology studies.
  • naturally derived saponin suitable for biochemical assays
  • reported D1 receptor binding for receptor pharmacology research
  • available in small, weighed quantities for analytical work
  • characterized by established identifiers such as CAS number for unambiguous referencing
  • intended for research use only; not for human or clinical use