Learn More
Cayman Chemical Bacopasaponin C, Purity: ≥95%, CAS: 178064-13-6, 10 mg
SDP
Catalog No. 502952675
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; inhibits the ATPase activity of P-glycoprotein (IC50 = 57.83 µg/ml); inhibits scopolamine-induced impairments in spatial memory in the Morris water maze and memory retrieval in the step-down test in mice at 50 mg/kg; has antidepressant-like activity in mice; reduces the spleen parasite load in a hamster model of leishmaniasis
Catalog No. 50-295-2675 Supplier Cayman Chemical Supplier No. 1182010 MG
A triterpenoid saponin with diverse biological activities; inhibits the ATPase activity of P-glycoprotein (IC50 = 57.83 µg/ml); inhibits scopolamine-induced impairments in spatial memory in the Morris water maze and memory retrieval in the step-down test in mice at 50 mg/kg; has antidepressant-like activity in mice; reduces the spleen parasite load in a hamster model of leishmaniasis