Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacoside A3, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013265 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3265 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3265 Supplier MilliporeSigma Supelco Supplier No. 538895MG
Add to Cart
Add to Cart

Bacoside A3 is a triterpenoid saponin and an important constituent of bacosides, a saponin mixture of Bacopa monniera.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 157408-08-7
Physical Form Neat
Linear Formula C47H76O18
MDL Number MFCD07779137
Synonym (3 beta,16 beta,23 R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-alpha-L-arabinofuranosyl-(1->2)-O-[beta-D-glucopyranosyl-(1->3)]-beta-D-glucopyranoside; Jujubogenin 3-O-[alpha-L-arabinofuranosyl-(1->2)-O-[beta-D-glucopyranosyl-(1->3)]-beta-D-glucopyranoside]
Type Analytical Standard
Quantity 5 mg
Formula Weight 929.10
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.