Learn More
Carolina™ Lactobacillus acidophilus, MicroKwik Culture™, Vial
SDPSCI_ED
Catalog No. s21820nd Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Vial
Catalog No. Quantity
S21820ND 1 Vial
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21820ND Supplier Carolina Biological Supply Supplier No. 155110A
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes
Note: Each MicroKwik Culture™ comes with 5mL of rehydration medium.
The culture is Gram+, nonmotile rods; used in production of acidophilus milk
Every culture is guaranteed for species and purity. Bacteria cultures are labeled by genus and species name, media on which they are cultivated, and temperature for optimal growth. All Gram staining is based on 24-hour cultures. Special packing and transportation costs are extra for animal and human pathogens.
  • Each culture contains enough material for a class of 30 students
  • It is cultured on tomato juice/yeast extract/milk broth at 37°C
  • MicroKwik Culture™ is a Carolina™ trademark for freeze-dried, viable bacteria plus medium
  • Since MicroKwik Cultures can be stored under refrigeration, they can be ordered ahead of time
Specifications
Quantity 1 Vial
Media Type Tomato Juice, Yeast Extract, Milk Broth
Product Type Bacteria Culture
Gram Stain Result Gram (+)