Learn More
Enzo Life Sciences AMPIPROBE Bacterial Vaginosis Extraction Controls (500µl)
SDP
Catalog No. 502013764
Encompass_Preferred
The AMPIPROBE Candida Extraction Controls are designed to be used during the genomic DNA extraction of Atopobium vaginae, Gardnerella vaginalis, Lactobacillus spp., Megasphaera spp., and BVAB2. All performance specifications must be validated by end-user and will vary based on chosen extraction methods. Eluent from extraction can be used as extraction control for AMPIPROBE Bacterial Vaginosis Assay kit (ENZ-GEN205). Plasmids in Extraction Controls are present at higher concentrations compared to assay controls included in the qPCR kit. Extra care should be taken to avoid contamination. Applications: qPCR. Species reactivity: BVAB-2, Megasphaera-1, Lactobacillus spp., Gardnerella vaginalis, Atopobium vaginae.
Catalog No. 50-201-3764 Supplier Enzo Life Sciences Supplier No. ENZGEN2060500
The AMPIPROBE Candida Extraction Controls are designed to be used during the genomic DNA extraction of Atopobium vaginae, Gardnerella vaginalis, Lactobacillus spp., Megasphaera spp., and BVAB2. All performance specifications must be validated by end-user and will vary based on chosen extraction methods. Eluent from extraction can be used as extraction control for AMPIPROBE Bacterial Vaginosis Assay kit (ENZ-GEN205). Plasmids in Extraction Controls are present at higher concentrations compared to assay controls included in the qPCR kit. Extra care should be taken to avoid contamination. Applications: qPCR. Species reactivity: BVAB-2, Megasphaera-1, Lactobacillus spp., Gardnerella vaginalis, Atopobium vaginae.