Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- BD BACTEC™ Myco F/Lytic Culture Vials are desgined for the recovery of mycobacteria from blood specimens and yeast and fungi from blood and sterile body fluids.
Specifications
Specifications
| Product Type | Blood Culture Media Vial |
| Target Organism Class | Mycobacteria |
| Test Type | Microbial Detection |
| Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?