Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- BD BACTEC™ Lytic Anaerobic medium is a medium designed to increase the detection and recovery of anaerobes.
- It contains a detergent to lyse both red and white blood cells present in the sample, releasing any intracellular organisms.
Specifications
Specifications
| Product Type | Blood Culture Media Vial |
| Target Organism Class | Aerobic Microorganisms |
| Test Type | Microbial Detection |
| Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?