Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BACTEC™ Platelet Anaerobic/F Culture Vials

Catalog No. B442054 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
B442054 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B442054 Supplier BD Supplier No. 442054
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A complete line of blood culture media for the detection of aerobes, anaerobes, facultative anaerobes, yeast, fungi and mycobacteria, all specifically developed to help improve time of detection and organism recovery.

  • BD BACTEC™ Platelet Anaerobic medium is formulated for the recovery of anaerobic microorganisms from leukocyte-reduced apheresis platelet (LRAP) units and leukocyte reduced whole blood platelet concentrates (LRWBPC).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Culture Media Vial
Target Organism Class Anaerobes
Test Type Microbial Detection
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.