Learn More
Enzo Life Sciences Bactenecin-5 precursor peptide (100µg)
SDP
Supplier:  Enzo Life Sciences BMLBW93150100
Encompass_Preferred
Bac5 is a 5-kDa proline-arginine rich antibiotic with a high degree of homology with PR39, an inhibitor of the 20S and 26S proteasome. Recently bactenecin 5 has been demonstrated to inhibit the chymotrypsin-like activity of the 20S proteasome. Alternative name: Bac5-GRR. Purity: ≥95% (HPLC). Solubility: Soluble in aqueous solution. Long Term Storage: -20°C.
Catalog No. 50-201-0340