Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bactenecin, G-Biosciences
SCI_ED

Catalog No. S162511
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162511 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S162511 Supplier G-Biosciences Supplier No. RC898
Add to Cart
Add to Cart

Bactenecin is a twelve residue antimicrobial peptide that is recognized by its disulfide linked cyclical structure.

It lyses gram-negative bacteria by increasing permeability through binding lipopolysaccharides (LPS). The bactenecin/LPS conjugate forms a Beta-hairpin structures which integrate with the bacterial membrane, forming pores. Cytosol contents leak with pore formation and lysis occurs with higher bactenecin concentrations

Specifications

Quantity 1 mg
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.