Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacteria Chart
SDPSCI_ED

Catalog No. S233715
Change view
Click to view available options
Contains 14 photomicrographs; Laminated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S233715 Contains 14 photomicrographs; Laminated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S233715 Supplier Biocam Communications Inc Supplier No. S233715

missing translation for 'validMainframeMsg'

Study excellent preparations of bacteria without a microscope.

  • Laminated chart contains 14 photomicrographs
  • Cocci, bacilli and spirilla bacterial forms are shown in full-color photomicrographs
  • Stained bacteria show minute detail
  • Photos of rods with flagella, spores, capsules, diplococcus, staphylococcus, spirochetes and many more

Specifications

Product Type Bacteria Chart
Includes 14 Photomicrographs, Cocci, bacilli and spirilla bacterial forms are shown, Stained bacteria, Photos of rods with flagella, spores, capsules, diplococcus, staphylococcus, spirochetes
For Use With (Application) Excellent preparations of bacteria without a microscope
Subject Science
Material Plastic, Laminated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.