Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bacteria protease native Protein from

Catalog No. 89958309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-309 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-309 Supplier Abnova™ Supplier No. P5339
Add to Cart
Add to Cart

Used for Enzyme, SDS-PAGE

Specifications

For Use With (Application) Enzyme Activity, SDS-PAGE
Name Protease (Staphylococcus aureus) Native Protein
Preparation Method Native protein isolated from the culture filtrate of Staphylococcus aureus, strain V8
Quantity 5 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name PROTEASE
Biological Activity ≥ 500 units/mg. Unit definition: One unit will change A280nm 0.001 per minute at 37°C, pH 7.8, using casein as the substrate.
Species Bacteria
Protein Tag None
Expression System Native protein isolated from the culture filtrate of Staphylococcus aureus, strain V8
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.