Learn More
Bioworld Bacteria Screening Medium 523, 500 g
Catalog No. 50254552
Encompass_Preferred
Bacteria Screening Medium 523Used to identify bacteria-free plant tissue for use in tissue culturepH: 5.5-6.5Dehydrated Appearance: yellow to cream powderSoluble in waterContains:Casein Hydrolysate Magnesium Sulphate Heptahydrate Potassium Phosphate Monobasic Yeast Extract Sucrose Agar
Catalog No. 50-254-552 Supplier Bioworld Supplier No. 306300021
Bacteria Screening Medium 523Used to identify bacteria-free plant tissue for use in tissue culturepH: 5.5-6.5Dehydrated Appearance: yellow to cream powderSoluble in waterContains:Casein Hydrolysate Magnesium Sulphate Heptahydrate Potassium Phosphate Monobasic Yeast Extract Sucrose Agar