Learn More
Biotium Bacteria Stains Fluorescence Microscopy Chemical Reagents Microbiology 1/EA
Catalog No. 5000501548
Encompass_Preferred
BactoSpore 488/540 Nucleic Acid Stain is a yellow fluorescent nucleic acid dye that stains live and dead gram-positive and gram-negative bacteria In B subtilis the dye stains both vegetative cells and endospores Ex/Em 488/536 nm (with DNA) 500X dye in DMSO 100 uL
Catalog No. 50-005-01548 Supplier Biotium Supplier No. 40120
BactoSpore 488/540 Nucleic Acid Stain is a yellow fluorescent nucleic acid dye that stains live and dead gram-positive and gram-negative bacteria In B subtilis the dye stains both vegetative cells and endospores Ex/Em 488/536 nm (with DNA) 500X dye in DMSO 100 uL