Learn More
Gold Biotechnology Inc Bacterial Cell Lysis Buffer
SDP
Catalog No. 5000562117
Encompass_Preferred
GoldBio s original formula for Bacterial Cell Lysis Buffer has been developed for the extraction of biologically active soluble proteins and inclusion bodies from bacterial cells The Bacterial Lysis Buffer is a proprietary improvement on the lysozyme based lysis method which allows extraction of soluble proteins and concurrent removal of nucleic acids (DNA RNA) released during cell lysis This lysis buffer eliminates viscosity build-up allowing effective clarification with lower centrifugal forces
Catalog No. 50-005-62117 Supplier Gold Biotechnology Inc Supplier No. GB177100
GoldBio s original formula for Bacterial Cell Lysis Buffer has been developed for the extraction of biologically active soluble proteins and inclusion bodies from bacterial cells The Bacterial Lysis Buffer is a proprietary improvement on the lysozyme based lysis method which allows extraction of soluble proteins and concurrent removal of nucleic acids (DNA RNA) released during cell lysis This lysis buffer eliminates viscosity build-up allowing effective clarification with lower centrifugal forces