Learn More
Electron Microscopy Sciences Bacterial Cell Spreader, Green
SDP
Catalog No. 501900436
Encompass_Preferred
Made from high impact polystyrene. Designed for easy spreading of cells onto the surface of an agar plate. Supplied sterile in individual packs with no flame sterilization needed. Total length of handle: 176 mm, width of spreader: 36 mm. Green.
Catalog No. 50-190-0436 Supplier Electron Microscopy Sciences Supplier No. 62452
Made from high impact polystyrene. Designed for easy spreading of cells onto the surface of an agar plate. Supplied sterile in individual packs with no flame sterilization needed. Total length of handle: 176 mm, width of spreader: 36 mm. Green.