Learn More
OYC AMERICAS Bacterial Protocatechuate 3,4-Dioxygenase, 300 units, > 3 Units/mg, 1 - 10°C, Solution

Supplier: OYC AMERICAS 46852004
This recombinant Bacterial Protocatechuate 3,4-Dioxygenase (rPCO) enzyme is engineered for sensitive and specific detection in *in vitro* diagnostic (IVD) assays. Sourced from *Acinetobacter baumannii* and produced in *E. coli*, it efficiently catalyzes the oxidative ring-cleavage of 3,4-dihydroxybenzoate to 3-carboxy-*cis,cis*-muconate. Its robust activity makes it valuable for applications requiring protocatechuate removal, enhancing the accuracy of diagnostic results.
- Enzyme name: Bacterial Protocatechuate 3,4-Dioxygenase (rPCO)
- Source: Recombinant from bacteria
- EC Number: 1.13.11.3
- Pack size: 300 units
- Specific activity: > 3 Units / mg
- Purity: Single major band on SDS-PAGE
- Contaminants: NADPH oxidase < 0.01 %, Alkaline phosphatase < 0.002 %
- Storage conditions: 1 - 10°C (do not freeze)
- Research use: For research use only (secondary)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.