Learn More
Medchemexpress LLC HY-P1883A 1mg Medchemexpress, Bacterial Sortase Substrate III, Abz/DNP (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1883A1MG
Encompass_Preferred
Medchemexpress, HY-P1883A 1mg Bacterial Sortase Substrate III, Abz/DNP (TFA) CAS: Bacterial Sortase Substrate III, Abz/DNP TFA is an internally quenched fluorescent peptide substrate. Staphylococcus aureus transpeptidase sortase A (SrtA) reacts with its native substrate Bacterial Sortase Substrate III, Abz/DNP, cleaving it and catalyzing the formation of an amide bond between the carboxyl group of threonine and the amino group of cell-wall crossbridges. Cleavage of this substrate can be monitored at Ex/Em=320 nm/420 nm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0544