Learn More
Alpco Ltd Bacterial Stool Panel I PCR Kit
SDP
Catalog No. NC2284585
Encompass
Bacterial Stool Panel I is a multiplex real-time PCR for the direct, qualitative detection and differentiation of STEC, Salmonella spp., Campylobacter spp. and EIEC/Shigella spp. in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284585 Supplier Alpco Ltd Supplier No. 86BS1HU100
Bacterial Stool Panel I is a multiplex real-time PCR for the direct, qualitative detection and differentiation of STEC, Salmonella spp., Campylobacter spp. and EIEC/Shigella spp. in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.