Learn More
Alpco Ltd Bacterial Stool Panel PCR Kit
SDP
Catalog No. NC2284597
Encompass
Bacterial Stool Panel is a multiplex real-time PCR for the direct, qualitative detection and differentiation of Campylobacter spp. (C. coli, C. lari, C. jejuni), Salmonella spp. and Yersinia enterocolitica in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284597 Supplier Alpco Ltd Supplier No. 86BSPHU100
Bacterial Stool Panel is a multiplex real-time PCR for the direct, qualitative detection and differentiation of Campylobacter spp. (C. coli, C. lari, C. jejuni), Salmonella spp. and Yersinia enterocolitica in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.