Learn More
Dojindo Molecular Technologies Inc -Bacstain- Bacterial Viability Detection Kit-DAPI/PI can be used to measure the ratio of membrane-damaged bacteria to total bacteria, multichannel evaluation for drug efficacy, and bacterial condition set
SDP
Catalog No. NC1933292
Encompass_Preferred
-Bacstain- Bacterial Viability Detection Kits are a series of products for fluorescent double-staining of bacteria. By combining different types of fluorescent stains, staining images can be acquired for each index (membrane impairment, respiratory activity, and esterase activity). Bacterial viability is generally assessed by colony formation in the nutrient-agar medium. However, this requires a long culturing time, and it is hard to recognize the growth of viable but nonculturable bacteria. -Bacstain- Bacterial Viability Detection Kit-DAPI/PI uses bacterial membrane damage as an index. DAPI permeates into bacteria to stain nucleic acids regardless of membrane damage. PI passes only through damaged bacterial membranes. This kit can be used to measure the ratio of membrane-damaged bacteria to total bacteria. Dojindo offers various reagents for bacteria research. (https://www.dojindo.com/observe-bacteria-from-various-parameters/)
Catalog No. NC1933292 Supplier Dojindo Molecular Technologies Inc Supplier No. BS0810
-Bacstain- Bacterial Viability Detection Kits are a series of products for fluorescent double-staining of bacteria. By combining different types of fluorescent stains, staining images can be acquired for each index (membrane impairment, respiratory activity, and esterase activity). Bacterial viability is generally assessed by colony formation in the nutrient-agar medium. However, this requires a long culturing time, and it is hard to recognize the growth of viable but nonculturable bacteria. -Bacstain- Bacterial Viability Detection Kit-DAPI/PI uses bacterial membrane damage as an index. DAPI permeates into bacteria to stain nucleic acids regardless of membrane damage. PI passes only through damaged bacterial membranes. This kit can be used to measure the ratio of membrane-damaged bacteria to total bacteria. Dojindo offers various reagents for bacteria research. (https://www.dojindo.com/observe-bacteria-from-various-parameters/)