Learn More
Andwin Scientific BACTERIOLOGICAL AGAR 500 G
SDP
Catalog No. NC0733856
Encompass_Preferred
The Bacteriological Agar is a solidification agent that can be used for the preparation of microbiological culture media. Most of microorganisms are not able to digest agar and thus making it an excellent solidifying agent. This product is acquired from certain species of marine red algae including Gelidium, Pterocladia, and Gracilaria. The agar has a white cream appearance and has a pH of 5.9 @ 20C. It is sold in the quantity of 500 grams.
Catalog No. NC0733856 Supplier Andwin Scientific Supplier No. A01102
The Bacteriological Agar is a solidification agent that can be used for the preparation of microbiological culture media. Most of microorganisms are not able to digest agar and thus making it an excellent solidifying agent. This product is acquired from certain species of marine red algae including Gelidium, Pterocladia, and Gracilaria. The agar has a white cream appearance and has a pH of 5.9 @ 20C. It is sold in the quantity of 500 grams.