Learn More
IBI SCIENTIFIC 3P BACTERIOLOGICAL AGAR 500G
Catalog No. NC0753539
Encompass_Preferred
Bacteriological Agar is a water soluble colloidal extract obtained from certain species of marine red algae; including Gelidium, Pterocladia, Gracilaria. Impurities, debris, minerals, and pigments are reduced to specific levels during manufacture.
Catalog No. NC0753539 Supplier IBI SCIENTIFIC 3P Supplier No. IB49170
Bacteriological Agar is a water soluble colloidal extract obtained from certain species of marine red algae; including Gelidium, Pterocladia, Gracilaria. Impurities, debris, minerals, and pigments are reduced to specific levels during manufacture.