Learn More
NEOGEN Bacteriological Peptone, 5 kg
SDP
Catalog No. 502015874
Encompass_Preferred
Bacteriological Peptone is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.An economical source of nutrients provided by a balanced mixture of meat peptones and tryptone. The growth requirements of most non-fastidious organisms will be fulfilled by the range of amino acids, peptides and proteoses in this mixture.For more information visit https://www.neogen.com/solutions/Microbiology/bacteriological-peptone
Catalog No. 50-201-5874 Supplier NEOGEN Supplier No. 700004741
Bacteriological Peptone is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.An economical source of nutrients provided by a balanced mixture of meat peptones and tryptone. The growth requirements of most non-fastidious organisms will be fulfilled by the range of amino acids, peptides and proteoses in this mixture.For more information visit https://www.neogen.com/solutions/Microbiology/bacteriological-peptone