Learn More
Med Vet International Bacteriostatic Sterile Water For Injection, 30mL
Catalog No. 502827793
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Bacteriostatic water (bacteriostatic water for injection) is sterile water containing 0.9% benzyl alcohol that is used to dilute or dissolve medications; the container can be reentered multiple times (usually by a sterile needle) and the benzyl alcohol suppresses or stops the growth of most potentially contaminating bacteria.
Catalog No. 50-282-7793 Supplier Med Vet International Supplier No. RXWATER30
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Bacteriostatic water (bacteriostatic water for injection) is sterile water containing 0.9% benzyl alcohol that is used to dilute or dissolve medications; the container can be reentered multiple times (usually by a sterile needle) and the benzyl alcohol suppresses or stops the growth of most potentially contaminating bacteria.