Learn More
BD Cell Analysis 3P Bacteriostatic Concentrate Solution, 10 L
Catalog No. NC0212357
Encompass
BD Accuri Bacteriostatic Concentrate Solution treats a total of 10L of Sheath. Flow cytometry (Routinely Tested). Regulatory status: For Research Use Only. Not for use in diagnostic or therapeutic procedures.
Catalog No. NC0212357 Supplier BD Cell Analysis 3P Supplier No. 653156
BD Accuri Bacteriostatic Concentrate Solution treats a total of 10L of Sheath. Flow cytometry (Routinely Tested). Regulatory status: For Research Use Only. Not for use in diagnostic or therapeutic procedures.