Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacteriostatic Water, Sterile, G-Biosciences
SCI_ED

Catalog No. S001123
Change view
Click to view available options
Quantity:
3 x 30 mL
5 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001123 5 x 10 mL
S001155 3 x 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001123 Supplier G-Biosciences Supplier No. 7862225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bacteriostatic water is sterile water containing 0.9% benzyl alcohol preservative, which prevents bacterial growth.

  • This solution is supplied in borosilicate glass vials for precise dilutions, featuring premium flip cap tops and self-healing butyl rubber stoppers for secure and repeated access.
  • It serves as an ideal solvent for dissolving chemicals.
  • Manufactured under strict quality control measures, it offers a dependable performance for various laboratory applications.
  • This solution is Research Use Only (RUO), not intended for medical use and is not approved for diagnostic or therapeutic applications or in neonates.

Chemical Identifiers

Molecular Weight (g/mol) 18.02

Specifications

Percent Purity >99%
Grade Laboratory
Sterility Sterile
Quantity 5 x 10 mL
Chemical Name or Material Benzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.