Learn More
Cayman Chemical Kocurin, Purity: ≥70%, CAS: 1374772-61-8, 1 mg
SDP
Catalog No. 502940565
Encompass_Preferred
A peptide antibiotic; active against MRSA (MIC = 0.25 µg/ml), as well as B. subtilis and E. faecium in a solid agar test at 8 µg/ml; active against E. faecium, E. faecalis, S. epidermidis, and clinical isolates of vancomycin-resistant enterococci (MICs = 0.004-1.025 µg/ml); increases survival in a mouse model of E. faecium-induced septicemia at 2.5, 5, and 10 mg/kg; decreases the number of CFUs in a mouse model of MRSA lung infection
Catalog No. 50-294-0565 Supplier Cayman Chemical Supplier No. 291251 MG
A peptide antibiotic; active against MRSA (MIC = 0.25 µg/ml), as well as B. subtilis and E. faecium in a solid agar test at 8 µg/ml; active against E. faecium, E. faecalis, S. epidermidis, and clinical isolates of vancomycin-resistant enterococci (MICs = 0.004-1.025 µg/ml); increases survival in a mouse model of E. faecium-induced septicemia at 2.5, 5, and 10 mg/kg; decreases the number of CFUs in a mouse model of MRSA lung infection